3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Catalog # MOLE90005195
Supplier: Molekula
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:614.89 g/mol
Merck Index:13,02052
Melting Pt:116 °C
Storage Temperature:Refrigerator
CAS Number:75621-03-3
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083