Order Entry
Finland
ContactUsLinkComponent
 
MULTISURE® HIV Rapid Test, MP Biomedicals
  43030-020
New Product
MULTISURE® HIV Rapid Test, MP Biomedicals
  43030-020
New Product

 

  • Description:
    HIV rapid test (CE), 20 tests
  • Species reactivity:
    Human
  • Storage temperature:
    2...28 °C
  • Pk:
    1 KIT

 

 

The MULTISURE ® HIV rapid test is intended as a rapid diagnostic test, developed to detect antibodies specific to HIV-1 gp120, HIV-1 gp41, HIV-1 p24 (also react with HIV-2) and HIV-2 gp36 antigens in human serum, plasma, finger pricked whole blood or whole blood with anti-coagulants.

  • Diagnostic platforms: Rapid test
  • Diagnostic solutions: Infectious disease
  • Disease screened: Human Immunodeficiency Virus (HIV)
  • Sample type: Plasma, serum, whole blood
  • Analyte/target: HIV 1 and 2 antibodies