Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Beakers
Measuring Cup, HDPE, Natural, Conical
Measuring Cup, HDPE, Natural, Conical
Measuring Cup, HDPE, Natural, Conical
Catalog # RIXI4-3000-0300
Supplier:
RIXIUS
New Product
Expand image
Measuring Cup, HDPE, Natural, Conical
Catalog # RIXI4-3000-0300
Supplier:
RIXIUS
New Product
Specifications
Capacity:
300 ml
Height:
94 mm
Pk:
891
Weight:
19 g
Specifications
About this item
Product Details
Product Details