Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microbiology Test Systems
Antibiotic Tests
Antibiotic assay discs, accessories for Neo-Sensitabs
Antibiotic assay discs, accessories for Neo-Sensitabs
Antibiotic assay discs, accessories for Neo-Sensitabs
Catalog # ROSC97105
Supplier:
ROSCO
CAS Number:
Expand image
Antibiotic assay discs, accessories for Neo-Sensitabs
Catalog # ROSC97105
Supplier:
ROSCO
CAS Number:
Specifications
Pk:
250
Description:
One cartridge single dispenser
Seq:
40
Specifications
About this item
Product Details
Product Details