Olmesartan medoxomil ≥97.5% (by HPLC)
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:144689-63-4
:
Formula:C₂₉H₃₀N₆O₆
MDL Number:MFCD00944911
:558.60 g/mol
Specification Test Results
- Appearance:White to almost white powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5%
- HPLC:≥97.5%
- Residue after ignition:≤0.1%