4-(trans-4-Pentylcyclohexyl)cyclohexanone
5 g
Catalog # TCIAP2435-5G
Supplier: TCI
4-(trans-4-Pentylcyclohexyl)cyclohexanone
Catalog # TCIAP2435-5G
Supplier: TCI
CAS Number: 84868-02-0
Jump to:
Product Details & Documents
- CAS Number:84868-02-0
- Formula:C₁₇H₃₀O
- MDL Number:MFCD04038798
- Molecular Weight:250.42 g/mol
Specifications
Specifications
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



