4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) fluorescent labeling reagent
Catalog # STMC75004
Supplier: STEMCELL TECHNOLOGIES
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:350.25 g/mol
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681