Showa DURACoil 546W Cut Resistant Gloves
Supplier: Showa
Certificates
About this item
Featuring an engineered DURACoil liner and reinforced with HPPE, this cut- and abrasion-resistant model is an ultra-comfortable, multi-use work glove.
- Cut-resistant, abrasion-resistant and oil-resistant
- Seamless knit
- Flexible and breathable
- Form-fitting
- Increased dexterity
- Machine washable
The foamed polyurethane palm coating provides good grip and breathability, and the light color makes it easy to identify soiling and contamination. A seamless and ergonomic design replicates the natural curves of the hand for enhanced dexterity and comfort over long periods of wear.
Certifications: Cat II. EN 388:2016+A1:2018, 4X42C.
Specifications
- Catalog No:
- BEST546W-9/XL
- BEST546W-8/L
- BEST546W-6/S
- BEST546W-7/M
- BEST546W-10/XXL
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile