Order Entry
Finland
ContactUsLinkComponent
McCartney Glass Bottles, Wide Mouth, With Aluminium Screw Cap
McCartney Glass Bottles, Wide Mouth, With Aluminium Screw Cap
Catalog # HIMEGW162-1X100NO
Supplier:  HIMEDIA
New Product
McCartney Glass Bottles, Wide Mouth, With Aluminium Screw Cap
Catalog # HIMEGW162-1X100NO
Supplier:  HIMEDIA
New Product

Specifications

  • Capacity:
    30 ml
  • Colour:
    Clear
  • Pk:
    100

Specifications

About this item

McCartney bottles are specially used to carry culture medium and bacterial growth can be done in these bottles.

  • Autoclavable
  • Made of neutral glass
  • Leak-proof clear glass bottle
  • With aluminium screw cap with rubber liner (grey bromo butyl rubber)
  • Size: 84×31 mm

These bottles along with the cap and rubber liner can be autoclaved easily. Also, these bottles can be sterilised with the medium present inside it.

Application: Chemical laboratory, biological laboratory, microbiological laboratory, diagnosis centres, industrial laboratory and various other laboratories.