Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
Hose Connector, PE-HD
Hose Connector, PE-HD
Hose Connector, PE-HD
Catalog # SEMA481
Supplier:
SEMADENI
New Product
Expand image
Hose Connector, PE-HD
Catalog # SEMA481
Supplier:
SEMADENI
New Product
Specifications
Ext.Ø:
15 mm
Description:
Hose connector, PE-HD, D = 9 mm
Pk:
1
Specifications
About this item
Product Details
Product Details