FAM Dye qPCR Calibration Solution *10000X*
AATB67032
New Product
:
- Pk:100 µl
- Description:FAM dye qPCR calibration solution *10000X*
FAM dye qPCR calibration solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
- Stable for 12 months
Calibration and verification should be run every six months or as needed.
: For research use only.