Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Chromatography Standards
Retention Time Mixture
Retention Time Mixture
Retention Time Mixture
Catalog # ACCUD-7169-RT-RI
Supplier:
AccuStandard
New Product
CAS Number:
Expand image
Retention Time Mixture
Catalog # ACCUD-7169-RT-RI
Supplier:
AccuStandard
New Product
CAS Number:
Specifications
Pk:
500 mg
Description:
Retention time mixture
Storage conditions:
Ambient (>5 °C)
Specifications
About this item
Product Details
Product Details