Anti-Solute carrier family 22 member 5 Rabbit Polyclonal Antibody
ANTIA100113-100
New Product
- Antibody type:Primary
- Antigen name:CDSP
- Antigen symbol:Solute carrier family 22 member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O76082
- Antigen synonyms:High-affinity sodium-dependent carnitine cotransporter|Solute carrier family 22 member 5|OCTN2|Solute carrier family 22 (organic cation/carnitine transporter) member 5|Organic cation/carnitine transporter 2|OCTN2VT|Slc22a5|S22A5_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:62 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC22A5 (amino acids 282-331)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to SLC22A5 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Solute carrier family 22 member 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human