Anti-CRISPR-Cas9 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Rabbit polyclonal antibody to CRISPR-Cas9 for WB, ICC/IF and IHC with samples derived from Staphylococcus aureus.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB: 1:1000 to 1:5000, ICC/IF: 1:5000, IHC: 1:500
Type: Primary
Antigen: CRISPR-Cas9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Staphylococcus aureus
Specifications
- Catalog No:
- ANTIA85318-50
- ANTIA85318-100
- Antigen symbol:
- CRISPR-Cas9
- CRISPR-Cas9
- Antigen name:
- Cas9
- Cas9
- Antigen synonyms:
- CRISPR-associated endonuclease Cas9/Csn1|SpyCas9|CRISPR-Cas9/Csn1|CRISPR-Cas9
- CRISPR-associated endonuclease Cas9/Csn1|SpyCas9|CRISPR-Cas9/Csn1|CRISPR-Cas9
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Staphylococcus aureus
- Staphylococcus aureus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# J7RUA5
- UniprotID# J7RUA5
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant fragment corresponding to amino acids 803-1053 of Staphylococcus aureus Cas9 (CCK74173), expressed in and purified from E. coli
- Recombinant fragment corresponding to amino acids 803-1053 of Staphylococcus aureus Cas9 (CCK74173), expressed in and purified from E. coli
- Form:
- Liquid
- Liquid
- Molecular weight:
- 124 kDa
- 124 kDa
- Storage buffer:
- Supplied as an aliquot of serum with 5 mM Sodium azide
- Supplied as an aliquot of serum with 5 mM Sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC