96-Well PCR Microplate Magnet
STMC100-1304
- Description:96-Well PCR microplate magnet
- Pk:1
The 96-Well PCR microplate magnet is designed for magnetic bead-based nucleic acid extraction and is compatible with EasySep™ Total nucleic acid extraction kit.
- Magnet for column-free nucleic acid extraction
This magnet has 24 round posts made of neodymium (NdFeB) magnet, generating a magnetic field to attract and separate bound nucleic acids in a 1-sided pellet. The aluminum magnet frame features a step-down edge and fits standard 96-well polymerase chain reaction (PCR) plates. The magnetic orientation is axial with the north poles up.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').