Order Entry
Finland
ContactUsLinkComponent
EasySep™ 'The Big Easy' Magnet
EasySep™ 'The Big Easy' Magnet
Catalog # STMC18001
undefined
EasySep™ 'The Big Easy' Magnet
Catalog # STMC18001
Specifications
  • Description:
    EasySep™ 'The Big Easy' magnet
  • Pk:
    1
Specifications
About this item
The EasySep™ 'The Big Easy' magnet is designed for cell separation procedures using EasySep™ reagents when processing a large number of cells, or when using RoboSep™.
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 14 ml (17×95 mm) polystyrene tube
'The Big Easy' magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ magnetic particles, without the use of columns.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').