Anti-cGAS (phospho Ser420) Rabbit Polyclonal Antibody
Catalog # ANTIA305847-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:C6orf150
- Antigen symbol:cGAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8C6L5
- Antigen synonyms:CGAS_HUMAN|cGAMP synthase|Chromosome 6 open reading frame 150|cGAS|Mab 21 domain containing 1|Cyclic GMP-AMP synthase|cGMP Synthase|Hypothetical protein LOC115004|h-cGAS
- Modification:phospho Ser420
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:62 kDa
- Sequence:FCSYH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around S420 of mouse CGAS (NP_775562.2).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to cGAS (phospho Ser420) for WB with samples derived from Human and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1000
Type: Primary
Antigen: cGAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat