Order Entry
Finland
ContactUsLinkComponent
 
Anti-cGAS Rabbit Polyclonal Antibody
Catalog # ANTIA16161-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-cGAS Rabbit Polyclonal Antibody
Catalog # ANTIA16161-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C6orf150
  • Antigen symbol:
    cGAS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8N884
  • Antigen synonyms:
    CGAS_HUMAN|cGAMP synthase|Chromosome 6 open reading frame 150|cGAS|Mab 21 domain containing 1|Cyclic GMP-AMP synthase|cGMP Synthase|Hypothetical protein LOC115004|h-cGAS
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    62 kDa
  • Sequence:
    KEEKCCRKDCLKLMKYLLEQLKERFKDKKHLDKFSSYHVKTAFFHVCTQNPQDSQWDRKDLGLCFDNCVTYFLQCLRTEKLENYFIPEFNLFSSNLIDKRSKEFLTKQIEYERNNEFPVFDEF
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 400-522 of human cGAS (NP_612450.2)
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification.
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to cGAS for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: cGAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat