Anti-VGluT1 Mouse Monoclonal Antibody [clone: S28-9]
Catalog # ANTIA304823-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BNPI
- Antigen symbol:VGluT1
- Clonality:Monoclonal
- Clone:S28-9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q62634
- Antigen synonyms:Slc17a7|Brain-specific Na(+)-dependent inorganic phosphate cotransporter|VGluT1|VGLU1_HUMAN|Brain specific Na dependent inorganic phosphate cotransporter|Brain specific Na (+) dependent inorganic phosphate cotransporter|Solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7|Solute carrier family 17 member 7|Vesicular glutamate transporter 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 493 - 560 (cytoplasmic C - terminus) of rat VGlut1.
- Tested applications:IHC
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [S28-9] antibody to VGluT1 for WB and IHC with samples derived from human, mouse and rat.
- Validated applications: WB, IHC
- Recommended dilutions: WB: 1:1000
Type: Primary
Antigen: VGluT1
Clonality: Monoclonal
Clone: S28-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat