Order Entry
Finland
ContactUsLinkComponent
Anti-ASIC1 Mouse Monoclonal Antibody [Clone: S271-44]
Anti-ASIC1 Mouse Monoclonal Antibody [Clone: S271-44]
  ANTIA305010-100
New Product
undefined
Anti-ASIC1 Mouse Monoclonal Antibody [Clone: S271-44]
  ANTIA305010-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ACCN2
  • Antigen symbol:
    ASIC1
  • Clonality:
    Monoclonal
  • Clone:
    S271-44
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q6NXK8
  • Antigen synonyms:
    brain sodium channel 2|BNaC2|amiloride-sensitive cation channel 2, neuronal|acid-sensing ion channel 1|acid-sensing ion channel 1a protein|ASIC1|ASIC|Cation channel, amiloride-sensitive, neuronal, 2|acid-sensing (proton-gated) ion channel 1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~60 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Fusion protein amino acids 460 - 526 (Cytoplasmic C - terminus) of mouse ASIC1 .
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [S271-44] antibody to ASIC1 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:1000

Type: Primary
Antigen: ASIC1
Clonality: Monoclonal
Clone: S271-44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat