Order Entry
Finland
Orders LinkContactUsLinkComponent
Mouse TIM 1 ELISA Kit

Mouse TIM 1 ELISA Kit

Mouse TIM 1 ELISA kit, 96 Tests
Catalog # ANTIA76850-96
Supplier:   Antibodies.com
Mouse TIM 1 ELISA Kit
Catalog # ANTIA76850-96
Supplier:  Antibodies.com
CAS Number:  
Mouse TIM 1 ELISA Kit
Mouse TIM 1 ELISA Kit

Mouse TIM 1 ELISA kit, 96 Tests

CAS Number:  
Price
565,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

Mouse TIM 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse TIM 1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 7,813 to 500 pg/ml
  • Sensitivity: 4,688 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse TIM 1 employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse TIM 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for TIM 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TIM 1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-TIM 1 Antibody, which binds the captured TIM 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of TIM 1 captured in each well. The concentration of TIM 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Mouse TIM 1 ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    7,813 - 500 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Mouse TIM 1 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse TIM 1. No significant cross-reactivity or interference between TIM 1 and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA76850-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    4,688 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    TIM 1
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.