Rabbit Complement C5a ELISA Kit
Rabbit Complement C5a ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit Complement C5a in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection range: 62,5 to 4000 pg/ml
- Sensitivity: 37,5 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%
Rabbit Complement C5a ELISA Kit (A75253) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rabbit Complement C5a in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C5a has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C5a present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement C5a Antibody, which binds the captured Complement C5a present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C5a captured in each well. The concentration of Complement C5a can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
- Pk:96 Tests
- Description:Rabbit complement C5a ELISA kit
- Primary antibody reactivity:Rabbit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:100 μl
- Detection range:62,5 - 4000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Rabbit Complement C5a ELISA Kit exhibits high specificity and excellent specificity for the detection of rabbit Complement C5a. No significant cross-reactivity or interference between Complement C5a and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:ANTIA75253-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:37,5 pg/ml
- Format:Pre-coated
- Target protein:Complement C5a
- Regulatory status:RUO



