Order Entry
Finland
Orders LinkContactUsLinkComponent
Human Complement C4a ELISA Kit

Human Complement C4a ELISA Kit

Human complement C4a ELISA kit, 96 Tests
Catalog # ANTIA79155-96
Supplier:   Antibodies.com
Human Complement C4a ELISA Kit
Catalog # ANTIA79155-96
Supplier:  Antibodies.com
CAS Number:  
Human Complement C4a ELISA Kit
Human Complement C4a ELISA Kit

Human complement C4a ELISA kit, 96 Tests

CAS Number:  
Price
625,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

Human Complement C4a ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Complement C4a in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 1,563 to 100 ng/ml
  • Sensitivity: 0,938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Complement C4a ELISA Kit (A79155) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Complement C4a in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C4a has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C4a present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement C4a Antibody, which binds the captured Complement C4a present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C4a captured in each well. The concentration of Complement C4a can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Human complement C4a ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    1,563 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Human Complement C4a ELISA Kit exhibits high specificity and excellent specificity for the detection of human Complement C4a. No significant cross-reactivity or interference between Complement C4a and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA79155-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,938 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Complement C4a
  • Regulatory status:
    RUO

Specifications