Mouse JNK3 ELISA Kit
Product Details & Documents
Mouse JNK3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse JNK3 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse JNK3 ELISA kit (A303465) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse JNK3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for JNK3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the JNK3 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-JNK3 Antibody, which binds the captured JNK3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of JNK3 captured in each well. The concentration of JNK3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- Pk:96 Tests
- Description:Mouse JNK3 ELISA kit
- Primary antibody reactivity:Mouse
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Mouse JNK3 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse JNK3. No significant cross-reactivity or interference between JNK3 and analogues was observed.
- Cat. No.:ANTIA303465-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:9,375 pg/ml
- Format:Pre-coated
- Target protein:JNK3
- Regulatory status:RUO



