Order Entry
Finland
Orders LinkContactUsLinkComponent
Human Cdk7 ELISA Kit

Human Cdk7 ELISA Kit

Human Cdk7 ELISA kit, 96 Tests
Catalog # ANTIA76295-96
Supplier:   Antibodies.com
Human Cdk7 ELISA Kit
Catalog # ANTIA76295-96
Supplier:  Antibodies.com
CAS Number:  
Human Cdk7 ELISA Kit
Human Cdk7 ELISA Kit

Human Cdk7 ELISA kit, 96 Tests

CAS Number:  
Price
625,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

Human Cdk7 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cdk7 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Cdk7 ELISA Kit (A76295) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Cdk7 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cdk7 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cdk7 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cdk7 Antibody, which binds the captured Cdk7 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cdk7 captured in each well. The concentration of Cdk7 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Human Cdk7 ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    0,156 - 10 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Human Cdk7 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Cdk7. No significant cross-reactivity or interference between Cdk7 and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA76295-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,094 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Cdk7
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.