Mouse PGP9.5 ELISA Kit
Product Details & Documents
Mouse PGP9.5 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PGP9.5 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 62,5 to 4000 pg/ml
- Sensitivity: 37,5 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse PGP9.5 ELISA Kit (A75937) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PGP9.5 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PGP9.5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PGP9.5 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PGP9.5 Antibody, which binds the captured PGP9.5 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PGP9.5 captured in each well. The concentration of PGP9.5 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- Pk:96 Tests
- Description:Mouse PGP9.5 ELISA kit
- Primary antibody reactivity:Mouse
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:100 μl
- Detection range:62,5 - 4000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Mouse PGP9.5 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse PGP9.5. No significant cross-reactivity or interference between PGP9.5 and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:ANTIA75937-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:37,5 pg/ml
- Format:Pre-coated
- Target protein:PGP9.5
- Regulatory status:RUO



