Centering Ring, KF25SVCR
EDWAKF25SVCR
New Product
- Description:Center ring used for vacuum hose connection to rotary pump
- Pk:1
Used for vacuum hose connection to rotary pump.
- Stainless steel with a black o-ring
- Compatiable with all Edwards and Leybold pump options