Order Entry
Finland
ContactUsLinkComponent
Glycerine, Hi-LR™
Glycerine, Hi-LR™
Catalog # HIMEGRM081-1L
Supplier:  HIMEDIA
New Product
   Safer Human and Environmental Health
CAS Number:  
Glycerine, Hi-LR™
Catalog # HIMEGRM081-1L
Supplier:  HIMEDIA
New Product
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)

Specifications

About this item

 :
 :92.09 g/mol
Storage Temperature:Room Temperature
EINECS:200-289-5
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722