Cumene
Catalog # ACSDSB224.500MG
Supplier: CPACHEM
New Product
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
Boiling Pt:152 °C
Melting Pt:−96,9 °C
Density:0,86 g/cm³
Flash Pt:36 °C
EINECS:202-704-5
UN:1918
ADR:3,III
:120.19 g/mol
:GHS02
:GHS08
:GHS09
:GHS07
CAS Number:98-82-8
Formula:C₉H₁₂
MDL Number:MFCD00008881
Melting Pt:–96,9 °C