Order Entry
Finland
ContactUsLinkComponent
Magicard™ Colour + Black Film for Prima 8, 750 Shot
Magicard™ Colour + Black Film for Prima 8, 750 Shot
Catalog # BRDY321978
Supplier:  Brady
New Product
undefined
Magicard™ Colour + Black Film for Prima 8, 750 Shot
Catalog # BRDY321978
Supplier:  Brady
New Product

Specifications

  • Colour:
    Assorted
  • Description:
    750 shot colour film with black on reverse, 4-colour ribbon (Cyan, Magenta, Yellow, Black), 750 single-sided cards
  • Length:
    7,31 m
  • Pk:
    1
  • Width:
    60 mm
  • For:
    Prima 8

Specifications

About this item

Prima 8 dye film allows full colour printing on one side and black on the other.

  • Print full colour on one side and black on the reverse side
  • Fits Prima 8 printers

Print 750 cards front and back in one go.

Delivery: Includes 1 dye film.