Anti-SLC6A18 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-82024
Supplier: Novus Biologicals
New Product
Specifications
- Antibody type:Primary
- Antigen name:Sodium- and chloride-dependent transporter XTRP2
- Antigen symbol:SLC6A18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Antigen synonyms:solute carrier family 6 (neurotransmitter transporter), member 18|FLJ31236|System B(0) neutral amino acid transporter AT3|Solute carrier family 6 member 18|XTRP2|solute carrier family 6, member 18|sodium-dependent neutral amino acid transporter B(0)AT3|sodium channel-like protein
- Purity:Immunogen affinity purified
- Storage buffer:PBS (pH 7,2) and 40% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids: GFKATNDYEHCLDRNILSLINDFDFPEQSISRDDYPAVLMHLNATWPKRVAQLPLKACLLEDFLDKSASGPGLAFVVFTETDL
- Pk:0,1 mL
Specifications
About this item
This antibody was developed against Recombinant Protein corresponding to amino acids: GFKATNDYEHCLDRNILSLINDFDFPEQSISRDDYPAVLMHLNATWPKRVAQLPLKACLLEDFLDKSASGPGLAFVVFTETDL
- For IHC-Paraffin, HIER pH 6 retrieval is recommended
Immunohistochemistry 1:500 - 1:1000
Immunohistochemistry-Paraffin 1:500 - 1:1000
Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Type: Primary
Antigen: SLC6A18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human