Order Entry
Finland
ContactUsLinkComponent
Human Recombinant IL-31
Human Recombinant IL-31
Certificates
undefined
Human Recombinant IL-31

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IL-31
  • Purity:
    ≥95%

Specifications