Order Entry
Finland
ContactUsLinkComponent
Rat Recombinant M-CSF
Rat Recombinant M-CSF
Catalog # STMC78117.1
CAS Number:  
undefined
Rat Recombinant M-CSF
Catalog # STMC78117.1
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Seq:
    1166
  • Protein/peptide name:
    M-CSF
  • Purity:
    ≥95%

Specifications

About this item

Macrophage colony-stimulating factor (M-CSF) is a homodimeric cytokine that belongs to receptor tyrosine kinase subclass III of the receptor tyrosine kinase (RTK) family. M-CSF acts on a CSF-1 receptor tyrosine kinase, which initiates signaling cascades to support cell proliferation and differentiation (Hamilton). M-CSF is produced by endothelial cells, osteoblasts, and during pregnancy by the uterine epithelial cells (Ryan). M-CSF synergizes with other factors to support proliferation and differentiation of multipotent hematopoietic progenitor cells, and regulates proliferation and differentiation of the mononuclear phagocyte progenitor cells to monocytes and macrophages. It also supports survival, proliferation, and function of the differentiated macrophages and regulates differentiation of mononuclear phagocytes to osteoclasts (Pixley and Stanley). M-CSF plays an important role in the implantation of the embryo and early development (Makrigiannakis et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').