Histopaque®, sterile filtered, Sigma-Aldrich®
Catalog # A7054-12X15ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Appearance (Turbidity):Clear
- Appearance (Form):Solution
- Appearance (Colour):Colourless
- Performance:Conforms
Specification Test Results
About this item
:
Density:1,077 g/ml
:M-Clarity™ quality level = MQ400
:GHS08
Storage Temperature:Refrigerator