Cibacron Blue 3G-A sodium salt ≥55% (dye content), Sigma-Aldrich®
B1064-1G
:
- Pk:1 g
- Seq:3047
- Appearance (Colour):Dark Blue
- Appearance (Form):Powder
- Solubility (Colour), 10 mg/ml, H2O:Dark Blue
- Nitrogen:≥7.0%
- Dye Content:≥55%
- Proton NMR Spectrum:Conforms to Structure
:
:840.11 g/mol
:M-Clarity™ quality level = MQ200
:Room Temperature
:84166-13-2
:C₂₉H₁₇CIN₇O₁₁S₃·3Na