Fast Blue RR Salt, capsules, Sigma-Aldrich®
Catalog # FBS25-10CAP
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:10
- Seq:0010
Specifications
Specification Test Results
- Appearance (Colour):Yellow and Yellow-Green
- Appearance (Form):Powder
- Performance, When staining with Sigma-Aldrich Procedure No. 85, cells stain in accordance with the package insert.:Conforms
Specification Test Results
About this item
:
:387.89 g/mol
:M-Clarity™ quality level = MQ500 (more info.)
Storage Temperature:Freezer
EINECS:238-777-5
CAS Number:14726-29-5
Formula:C₁₅H₁₄ClN₃O₃
MDL Number:MFCD00074762