Bromobimane (mBBr) ≥97%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:71418-44-5
:
Formula:C₁₀H₁₁BrN₂O₂
MDL Number:MFCD00036951
:271.11 g/mol
Melting Pt:161 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
Specification Test Results
- Appearance (Colour):Yellow
- Appearance (Form):Powder
- Solubility (Colour):Yellow-Green
- Solubility (Turbidity), 20 mg/ml, Acetonitrile:Clear
- Proton NMR spectrum:Conforms to Structure
- EmM in H2O:4.6 - 5.1
- Wavelength:396 - 398 nm
- % Purity (HPLC):≥97