Order Entry
Finland
ContactUsLinkComponent
Histopaque®, sterile filtered, Sigma-Aldrich®
Histopaque®, sterile filtered, Sigma-Aldrich®
Catalog # A0561-100X15ML
CAS Number:  
Histopaque®, sterile filtered, Sigma-Aldrich®
Catalog # A0561-100X15ML
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Appearance (Turbidity):
    Clear
  • Appearance (Form):
    Solution
  • Appearance (Colour):
    Colorless
  • Performance:
    Conforms

Specification Test Results

About this item

 :
Density:1,077 g/ml
 :M-Clarity™ quality level = MQ400
 :GHS08
Storage Temperature:Refrigerator