Order Entry
Finland
ContactUsLinkComponent
Brilliant cresyl blue
Brilliant cresyl blue
Catalog # MOLE45545810
Supplier:  Molekula
CAS Number:  
Brilliant cresyl blue
Catalog # MOLE45545810
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :801.96 g/mol
EINECS:279-675-0
CAS Number:81029-05-2
 :10127-36-3
Formula:C₃₄H₄₂Cl₄N₈O₂Zn
MDL Number:MFCD00148901