Order Entry
Finland
ContactUsLinkComponent
Brilliant cresyl blue, pure, high purity biological stain
Brilliant cresyl blue, pure, high purity biological stain
Certificates
undefined
Brilliant cresyl blue, pure, high purity biological stain

Specification Test Results

  • Appearance:
    Green to dark blue Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤8 % (1 g, 110°C)
  • Solubility:
    (0.1% in 50% ethanol) Clear blue
  • Specific absorbance A (1%/1cm):
    ≥1500 (625 to 630 nm, 50% ethanol)

Specification Test Results

Related Information