SteriWare® Scrapers, Blue
Supplier: Sampling Systems
Certificates
About this item
The SteriWare scraper is designed for use in production or laboratory applications.
- Available in non-sterile or sterile formats
- 75 mm blade width
- Used for scraping, removing and splitting product
- Available from stock
- Fast delivery
These blue disposable scrapers are ideal for use in multiple applications, such as removing excess product or scraping machines clean after use. The bright blue colour allows it to be easily spotted if left in material.