Tight Head Drums
Supplier: Sampling Systems
Certificates
About this item
Tight head drums are perfect for use in food, chemical, pharmaceutical, and cosmetic manufacturing applications.
- Made from high quality 316L stainless steel
- Fitted with a 2" and ³/₄" bung
- Comes with an EPDM seal
- Available from stock
- Fast delivery
High-quality stainless steel, closed top drums are ideal for use across several industries, including pharma and food. They area available in a wide range of sizes and have swaged sides for easier handling and added strength.