Chromazurol B
Catalog # M1782-1G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Brown with an orange to red cast powder
- Solubility:Slightly hazy orange solution at 4 mg plus 4 ml of water
- Dye content:approx. 60%
- undefined:Approved KWK 9/5/96
Specification Test Results
About this item
:
:503.24 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
CAS Number:1796-92-5
Formula:C₂₃H₁₄Cl₂Na₂O₆
MDL Number:MFCD00036446