ImmunoHistoMount™ Mounting Medium
Catalog # I1161-30ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:30 mL
- M-Clarity quality level:MQ100
- Storage temperature:RT
- pH:7.6 - 8.0
- Application:Hematology, histology
Specifications
About this item
ImmunoHistoMount™ is a permanent aqueous mounting medium designed for tissue sections and cell smears with peroxidase and alkaline phosphatase chromogens that cannot be dehydrated with organic solvents.
- Aqueous-based mounting medium comparable to Crystal Mount™
- Can be used in hematology and histology applications
This mounting medium preserves fast red, aminoethylycarbazole (AEC), BCIP/NBT, and BCIP/INT chromogens and is compatible with counterstains like hematoxylin and nuclear fast red (NFR). ImmunoHistoMount is also suitable for chromogens like DAB and DAB with nickel and cobalt. It is not compatible with routine H & E staining.