Calcein blue fluorescent dye, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:54375-47-2
:
Formula:C₁₅H₁₅NO₇
MDL Number:MFCD00006867
:321.29 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):White to Light Yellow
- Appearance (Form):Powder
- Solubility (Colour):Light Yellow to Dark Yellow and Light Orange to Dark Orange
- Solubility (Turbidity)
50 mg/mL, 1 M NaOH:Clear to Slightly Hazy - Carbon:54.5 - 56.5 %
- Nitrogen:4.0 - 4.6 %
- Suitability
for use as an indicator for EDTA titrations:Suitable - 1H NMR Spectrum:Conforms to Structure