Silver methenamine
Catalog # HT1003-6VL
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Vial of 110 mg
- Pk:6
- Seq:0006
Specifications
Specification Test Results
- Appearance (Colour):White to Light Grey
- Appearance (Form):Powder
- Performance:Conforms
- 24 months:When using Sigma-Aldrich Procedure No. HT100, fungi organisms stain purplish-brown to black
- undefined:Expiration Date Period
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ500 (more info.)
:248.06 g/mol
:GHS05
:GHS07
:GHS08
Storage Temperature:Refrigerator
CAS Number:37604-90-3
Formula:C₆H₁₂AgN₄
MDL Number:MFCD00284609