Haematoxylin solution Gill III, Sigma-Aldrich®
Catalog # GHS380-2.5L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Appearance (Colour)
Dark Reddish-Purple:Conforms - Appearance (Form):Solution
- Appearance (Turbidity)
Essentially free of precipitate:Clear - Performance:Conforms
- Expiration Date Period:24 months
Specification Test Results
About this item
:
:302.28 g/mol
:M-Clarity™ quality level = MQ500 (more info.)
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Room Temperature
CAS Number:517-28-2
:208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111