Order Entry
Finland
ContactUsLinkComponent
 
Giemsa's stain, Sigma-Aldrich®
Catalog # GS128-4L
CAS Number:  
undefined
Giemsa's stain, Sigma-Aldrich®
Catalog # GS128-4L
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Appearance (Turbidity):
    Clear
  • Appearance (Colour):
    Faint Blue to Dark Blue
  • Appearance (Form):
    Solution
  • pH
    at 25 degrees C:
    6.75 - 6.95
  • Performance:
    Conforms
  • Expiration Date Period:
    24 months

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
Flash Pt:11 °C (closed cup)
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Room Temperature
CAS Number:51811-82-6
MDL Number:MFCD00081642