Anti-CD2 Recombinant Antibody [Clone: YTH 655]
Catalog # PRSI24-191
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD2 molecule
- Antigen symbol:CD2
- Clonality:Recombinant
- Clone:YTH 655
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:914
- Antigen synonyms:Rosette receptor|Erythrocyte receptor|T-cell surface antigen T11/Leu-5|T-cell surface antigen CD2|LFA-3 receptor|LFA-2
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Tested applications:In vivo Depletion
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD2 molecule
Clonality: Recombinant
Clone: YTH 655
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human