Order Entry
Finland
ContactUsLinkComponent
Anti-Il4 Recombinant Antibody [Clone: 11B11]
Anti-Il4 Recombinant Antibody [Clone: 11B11]
Catalog # PRSI24-185
Supplier:  ProSci Inc.
New Product
Anti-Il4 Recombinant Antibody [Clone: 11B11]
Catalog # PRSI24-185
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    interleukin 4
  • Antigen symbol:
    Il4
  • Clonality:
    Recombinant
  • Clone:
    11B11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    16189
  • Antigen synonyms:
    B-cell stimulatory factor 1|B-cell growth factor 1|Il4|B-cell IgG differentiation factor|Il-4|BSF-1|Interleukin-4
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    BSF-1 / IL-4 purified from serum-free supernatants of cultured EL-4 cells (a C57BL/6 thymoma) induced with 4β-phorbol 12β-myristate 12α-acetate for 48h.
  • Tested applications:
    in vivo
  • Size:
    0,2 mg
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: interleukin 4
Clonality: Recombinant
Clone: 11B11
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse